Product Details

SNP ID
rs140309148
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37470024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATAGAGCAGACAATGGAGGGAG[C/T]GAAATCTGCAGAGAGACCAGAAAAG
Phenotype
MIM: 602577
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MFNG PubMed Links

Gene Details

Gene
MFNG
Gene Name
MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166343.1 1073 Missense Mutation CAC,CGC H288R NP_001159815.1
NM_002405.3 1073 Missense Mutation CAC,CGC H302R NP_002396.2

View Full Product Details