Product Details

SNP ID
rs140711281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29308534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTGGTGCTGTGCCTGCTGGAGGT[A/G]GCGCGGCGTGGGGCACGCCTGGGCC
Phenotype
MIM: 133450 MIM: 602128 MIM: 602220
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EWSR1 PubMed Links

Gene Details

Gene
EWSR1
Gene Name
EWS RNA binding protein 1
There are no transcripts associated with this gene.

Gene
GAS2L1
Gene Name
growth arrest specific 2 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278730.1 607 Silent Mutation GTA,GTG V143V NP_001265659.1
NM_006478.4 607 Silent Mutation GTA,GTG V143V NP_006469.2
NM_152236.2 607 Intron NP_689422.1
XM_011529824.1 607 Silent Mutation GTA,GTG V143V XP_011528126.1
XM_011529825.1 607 Silent Mutation GTA,GTG V143V XP_011528127.1
XM_017028533.1 607 Silent Mutation GTA,GTG V143V XP_016884022.1
Gene
RASL10A
Gene Name
RAS like family 10 member A
There are no transcripts associated with this gene.

View Full Product Details