Product Details

SNP ID
rs140738257
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:17738149 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCAGCAGCAGGGCCAGCACCAG[C/T]ATGGTCTTCTCCTTCTCCATGTCTC
Phenotype
MIM: 601997
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCL2L13 PubMed Links

Gene Details

Gene
BCL2L13
Gene Name
BCL2 like 13
There are no transcripts associated with this gene.

Gene
BID
Gene Name
BH3 interacting domain death agonist
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001196.3 547 Missense Mutation ATA,ATG I148M NP_001187.1
NM_001244567.1 547 Missense Mutation ATA,ATG I148M NP_001231496.1
NM_001244569.1 547 Missense Mutation ATA,ATG I52M NP_001231498.1
NM_001244570.1 547 Missense Mutation ATA,ATG I52M NP_001231499.1
NM_001244572.1 547 Missense Mutation ATA,ATG I52M NP_001231501.1
NM_197966.2 547 Missense Mutation ATA,ATG I194M NP_932070.1
NM_197967.2 547 Missense Mutation ATA,ATG I52M NP_932071.1
XM_017028906.1 547 Missense Mutation ATA,ATG I148M XP_016884395.1

View Full Product Details