Product Details

SNP ID
rs141025723
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20315469 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCTGCTCCTCACGGATCCGGTGT[C/T]CCACGGCCTGCCATGGGGCCAGGGC
Phenotype
MIM: 609459
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGCR6L PubMed Links

Gene Details

Gene
DGCR6L
Gene Name
DiGeorge syndrome critical region gene 6-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033257.3 Intron NP_150282.2
Gene
LOC440792
Gene Name
proline dehydrogenase 1 pseudogene
There are no transcripts associated with this gene.

View Full Product Details