Product Details

SNP ID
rs141274365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41925424 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCAGTTCAGTGGAGCCCAGCTC[C/T]GTGGCTGGCACAGGGACACTGTGGC
Phenotype
MIM: 610152 MIM: 606269
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CENPM PubMed Links

Gene Details

Gene
CENPM
Gene Name
centromere protein M
There are no transcripts associated with this gene.

Gene
MIR378I
Gene Name
microRNA 378i
There are no transcripts associated with this gene.

Gene
TNFRSF13C
Gene Name
TNF receptor superfamily member 13C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052945.3 542 Silent Mutation ACA,ACG T166T NP_443177.1

View Full Product Details