Product Details

SNP ID
rs141324912
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:32218662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCCTTGGTTAGCTTGGGTCCC[G/T]TCTGCAAACCGCAGAACAAGTCATA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC5A4 PubMed Links

Gene Details

Gene
SLC5A4
Gene Name
solute carrier family 5 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014227.2 1714 Missense Mutation NP_055042.1
XM_006724308.3 1714 Missense Mutation XP_006724371.1
XM_011530342.2 1714 Missense Mutation XP_011528644.1
XM_011530343.2 1714 Missense Mutation XP_011528645.1
XM_011530344.2 1714 Missense Mutation XP_011528646.1
XM_017028920.1 1714 Missense Mutation XP_016884409.1

View Full Product Details