Product Details

SNP ID
rs141942820
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:25354709 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCCGCTTGATGCTGTGGTGCTGC[C/G]AGGCGGTCCAGTAGATGAAGTCCCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC100652901 PubMed Links

Gene Details

Gene
LOC100652901
Gene Name
uncharacterized LOC100652901
There are no transcripts associated with this gene.

Gene
LRP5L
Gene Name
LDL receptor related protein 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135772.1 3668 Missense Mutation TCG,TGG S181W NP_001129244.1
NM_182492.2 3668 Intron NP_872298.1
XM_006724372.3 3668 Missense Mutation TCG,TGG S181W XP_006724435.1
XM_017029083.1 3668 Missense Mutation TCG,TGG S181W XP_016884572.1
XM_017029084.1 3668 Missense Mutation TCG,TGG S181W XP_016884573.1
XM_017029085.1 3668 Missense Mutation TCG,TGG S181W XP_016884574.1
XM_017029086.1 3668 Missense Mutation TCG,TGG S181W XP_016884575.1
XM_017029087.1 3668 Missense Mutation TCG,TGG S181W XP_016884576.1
XM_017029088.1 3668 Missense Mutation TCG,TGG S181W XP_016884577.1

View Full Product Details