Product Details

SNP ID
rs142028345
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37665630 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTGGTGGGCAAGCCCAGCCCCT[A/T]CATGTTCGAGTGCATCACGGAGAAC
Phenotype
MIM: 150570 MIM: 609246
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LGALS1 PubMed Links

Gene Details

Gene
LGALS1
Gene Name
galectin 1
There are no transcripts associated with this gene.

Gene
LOC101927051
Gene Name
uncharacterized LOC101927051
There are no transcripts associated with this gene.

Gene
PDXP
Gene Name
pyridoxal phosphatase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020315.4 703 Missense Mutation TAC,TTC Y217F NP_064711.1
Gene
SH3BP1
Gene Name
SH3 domain binding protein 1
There are no transcripts associated with this gene.

View Full Product Details