Product Details

SNP ID
rs142219748
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42516123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGAGGCCACAGGTGGACAGGAGG[C/T]GGGACAGGCTCTCCTGCCGCAGGGA
Phenotype
MIM: 607979
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RRP7A PubMed Links

Gene Details

Gene
RRP7A
Gene Name
ribosomal RNA processing 7 homolog A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015703.4 267 Missense Mutation CAC,CGC H77R NP_056518.2
Gene
SERHL
Gene Name
serine hydrolase-like (pseudogene)
There are no transcripts associated with this gene.

View Full Product Details