Product Details

SNP ID
rs142283864
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:37570312 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGTTGAACCCGCCCCTTACGCTC[A/G]GGTAGCTCAGGTGGCTGTGACCCAG
Phenotype
MIM: 606084 MIM: 150571
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDC42EP1 PubMed Links

Gene Details

Gene
CDC42EP1
Gene Name
CDC42 effector protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152243.2 462 Intron NP_689449.1
XM_006724107.1 462 Intron XP_006724170.1
XM_006724108.1 462 Intron XP_006724171.1
XM_006724109.1 462 Intron XP_006724172.1
XM_017028541.1 462 Intron XP_016884030.1
Gene
LGALS2
Gene Name
galectin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006498.2 462 Missense Mutation CCG,CTG P117L NP_006489.1

View Full Product Details