Product Details

SNP ID
rs142545364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31440093 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTCTGAACGCTGACAGCTGCTGC[A/G]TGGGAACCAGAGCCTGGAGGATGCA
Phenotype
MIM: 603952 MIM: 607445
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DRG1 PubMed Links

Gene Details

Gene
DRG1
Gene Name
developmentally regulated GTP binding protein 1
There are no transcripts associated with this gene.

Gene
EIF4ENIF1
Gene Name
eukaryotic translation initiation factor 4E nuclear import factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164501.1 3247 Silent Mutation NP_001157973.1
NM_001164502.1 3247 Silent Mutation NP_001157974.1
NM_019843.3 3247 Silent Mutation NP_062817.2
XM_005261686.2 3247 Missense Mutation XP_005261743.1
XM_005261687.2 3247 Missense Mutation XP_005261744.1
XM_005261688.2 3247 Missense Mutation XP_005261745.1
XM_006724281.3 3247 Missense Mutation XP_006724344.3
XM_011530280.1 3247 Silent Mutation XP_011528582.1
XM_011530281.2 3247 Missense Mutation XP_011528583.1
XM_017028863.1 3247 Missense Mutation XP_016884352.1
XM_017028864.1 3247 Missense Mutation XP_016884353.1

View Full Product Details