Product Details

SNP ID
rs142823248
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50512850 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAGCCATTGCACTGGCCAGAAT[A/G]TTCTTCTAGTACTGTCATGATCCTA
Phenotype
MIM: 611230
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LMF2 PubMed Links

Gene Details

Gene
LMF2
Gene Name
lipase maturation factor 2
There are no transcripts associated with this gene.

Gene
NCAPH2
Gene Name
non-SMC condensin II complex subunit H2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185011.1 Intron NP_001171940.1
NM_014551.4 Intron NP_055366.3
NM_152299.3 Intron NP_689512.2
XM_005261912.4 Intron XP_005261969.1
XM_011530685.2 Intron XP_011528987.1
XM_017028793.1 Intron XP_016884282.1

View Full Product Details