Product Details

SNP ID
rs142936184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:18913501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCCAAAGTACACCCGGTGGTCAG[C/T]AGGATGCAGGCCCAGCTCCTCCATC
Phenotype
MIM: 601279 MIM: 606810
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGCR6 PubMed Links

Gene Details

Gene
DGCR6
Gene Name
DiGeorge syndrome critical region gene 6
There are no transcripts associated with this gene.

Gene
PRODH
Gene Name
proline dehydrogenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195226.1 Intron NP_001182155.1
NM_016335.4 Intron NP_057419.4

View Full Product Details