Product Details

SNP ID
rs143430039
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38298813 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCAGCATGTTCCAGTCAAAGAC[A/G]TAGTCATAGGAGAAGCCCTGCCGGT
Phenotype
MIM: 600863
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CSNK1E PubMed Links

Gene Details

Gene
CSNK1E
Gene Name
casein kinase 1 epsilon
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001894.4 1324 Silent Mutation TAC,TAT Y286Y NP_001885.1
NM_152221.2 1324 Silent Mutation TAC,TAT Y286Y NP_689407.1
Gene
LOC400927-CSNK1E
Gene Name
LOC400927-CSNK1E readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289912.1 1324 Silent Mutation TAC,TAT Y286Y NP_001276841.1

View Full Product Details