Product Details

SNP ID
rs143483218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30379804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAACTCGTGCTTACAGGGCAGCA[A/C]CCGGAGCCACTACAGGGGTGGGGGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCDC157 PubMed Links

Gene Details

Gene
CCDC157
Gene Name
coiled-coil domain containing 157
There are no transcripts associated with this gene.

Gene
KIAA1656
Gene Name
KIAA1656 protein
There are no transcripts associated with this gene.

Gene
RNF215
Gene Name
ring finger protein 215
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017981.1 1018 Missense Mutation GTG,TTG V340L NP_001017981.1

View Full Product Details