Product Details

SNP ID
rs143929366
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30243911 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTTCTGGTCCCGGGTGATGTTGC[C/T]CAGGGAGGTGCCAAGGTACACGACT
Phenotype
MIM: 159540
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
LIF PubMed Links

Gene Details

Gene
LIF
Gene Name
leukemia inhibitory factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257135.1 505 Missense Mutation GAG,GGG E57G NP_001244064.1
NM_002309.4 505 Missense Mutation AGC,GGC S117G NP_002300.1
Gene
MGC20647
Gene Name
uncharacterized protein MGC20647
There are no transcripts associated with this gene.

View Full Product Details