Product Details
- SNP ID
-
rs144494015
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:31326898 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGACACAGCAGCTGCCTCATTTC[C/T]CTTCAGGCCCCATGGGCTGCTGGTC
- Phenotype
-
MIM: 605165
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PATZ1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2240424] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PATZ1
- Gene Name
- POZ/BTB and AT hook containing zinc finger 1
- Gene
- PIK3IP1-AS1
- Gene Name
- PIK3IP1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.
View Full Product Details