Product Details

SNP ID
rs144771084
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42060373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCACTGTGAAGTTGGTTTCATCT[C/T]GGAGGCCACTGATGATGTCACCTGA
Phenotype
MIM: 104170 MIM: 610981
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAGA PubMed Links

Gene Details

Gene
NAGA
Gene Name
alpha-N-acetylgalactosaminidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000262.2 1687 Missense Mutation CAA,CGA Q381R NP_000253.1
XM_005261615.4 1687 Missense Mutation CAA,CGA Q381R XP_005261672.1
XM_005261616.4 1687 Missense Mutation CAA,CGA Q381R XP_005261673.1
Gene
WBP2NL
Gene Name
WBP2 N-terminal like
There are no transcripts associated with this gene.

View Full Product Details