Product Details
- SNP ID
-
rs144824689
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:42693081 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTCCTCAAAGTACTTCTTCCAGT[A/C]CTGCCAGGGGATGGGGTAGAAGGCC
- Phenotype
-
MIM: 607922
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
A4GALT
PubMed Links
Gene Details
- Gene
- A4GALT
- Gene Name
- alpha 1,4-galactosyltransferase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001318038.1 |
1359 |
Missense Mutation |
GAC,TAC |
D291Y |
NP_001304967.1 |
NM_017436.5 |
1359 |
Missense Mutation |
GAC,TAC |
D291Y |
NP_059132.1 |
XM_005261644.2 |
1359 |
Missense Mutation |
GAC,TAC |
D291Y |
XP_005261701.1 |
XM_005261646.4 |
1359 |
Missense Mutation |
GAC,TAC |
D291Y |
XP_005261703.1 |
XM_005261647.2 |
1359 |
Missense Mutation |
GAC,TAC |
D291Y |
XP_005261704.1 |
XM_005261648.3 |
1359 |
Missense Mutation |
GAC,TAC |
D291Y |
XP_005261705.1 |
XM_006724265.3 |
1359 |
Missense Mutation |
GAC,TAC |
D291Y |
XP_006724328.1 |
XM_006724266.3 |
1359 |
Missense Mutation |
GAC,TAC |
D291Y |
XP_006724329.1 |
XM_011530233.2 |
1359 |
Missense Mutation |
GAC,TAC |
D291Y |
XP_011528535.1 |
XM_017028831.1 |
1359 |
Missense Mutation |
GAC,TAC |
D291Y |
XP_016884320.1 |
View Full Product Details