Product Details

SNP ID
rs144963144
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36257357 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCAAGTGGGACAGATACTGGAG[A/G]TCCTCAAAGTAAGCCCCTCGGTGAC
Phenotype
MIM: 603743
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
APOL1 PubMed Links

Gene Details

Gene
APOL1
Gene Name
apolipoprotein L1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136540.1 Intron NP_001130012.1
NM_001136541.1 Intron NP_001130013.1
NM_003661.3 Intron NP_003652.2
NM_145343.2 Intron NP_663318.1
XM_005261796.3 Intron XP_005261853.1
XM_011530478.2 Intron XP_011528780.1

View Full Product Details