Product Details

SNP ID
rs145150314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:22487965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAACATTTGATGACACTGGGTCTT[A/G]TGCTCCATTTTCTCCTTGAAAGTTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF280B PubMed Links

Gene Details

Gene
ZNF280B
Gene Name
zinc finger protein 280B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080764.3 1843 Silent Mutation NP_542942.2
XM_011529893.2 1843 Silent Mutation XP_011528195.1
XM_011529895.2 1843 Missense Mutation XP_011528197.1
XM_011529896.2 1843 Silent Mutation XP_011528198.1
XM_011529897.2 1843 Silent Mutation XP_011528199.1

View Full Product Details