Product Details

SNP ID
rs145411434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38484618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATGTCCCTATCCTGTTGTAGTG[A/T]ACACAATAGCAGAAAATTCTTTCTG
Phenotype
MIM: 608469
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DDX17 PubMed Links

Gene Details

Gene
DDX17
Gene Name
DEAD-box helicase 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098504.1 3616 UTR 3 NP_001091974.1
NM_006386.4 3616 UTR 3 NP_006377.2
Gene
KDELR3
Gene Name
KDEL endoplasmic reticulum protein retention receptor 3
There are no transcripts associated with this gene.

View Full Product Details