Product Details

SNP ID
rs145653843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31216609 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAATAAAAGATGCCTCCTGACAT[A/G]GGCAGGCCAGGGTGACTTGGATGGT
Phenotype
MIM: 601988
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LIMK2 PubMed Links

Gene Details

Gene
LIMK2
Gene Name
LIM domain kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031801.1 Intron NP_001026971.1
NM_005569.3 Intron NP_005560.1
NM_016733.2 Intron NP_057952.1
Gene
RNF185
Gene Name
ring finger protein 185
There are no transcripts associated with this gene.

View Full Product Details