Product Details

SNP ID
rs145805898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:49998195 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACGGCAGCCGGCCCCCGGCCCC[A/G]GGCGCCAGCACAGGCTCACATGGCC
Phenotype
MIM: 613414
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL17REL PubMed Links

Gene Details

Gene
IL17REL
Gene Name
interleukin 17 receptor E like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001694.2 2113 Missense Mutation CCG,CTG P239L NP_001001694.2
XM_011530689.2 2113 Missense Mutation CCG,CTG P311L XP_011528991.1
XM_017028798.1 2113 Missense Mutation CCG,CTG P311L XP_016884287.1

View Full Product Details