Product Details

SNP ID
rs146237824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50177693 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTCATCTTCGACAAGCTGCACAA[A/G]GTGGGCATCAAGACGCGCCGGCAGT
Phenotype
MIM: 608421
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PANX2 PubMed Links

Gene Details

Gene
PANX2
Gene Name
pannexin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160300.1 981 Silent Mutation AAA,AAG K327K NP_001153772.1
NM_052839.3 981 Silent Mutation AAA,AAG K327K NP_443071.2
Gene
TRABD
Gene Name
TraB domain containing
There are no transcripts associated with this gene.

View Full Product Details