Product Details

SNP ID
rs146609012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36991519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCCTGCCATGCCAGTGTTCTAAG[C/T]CCCTCACTGATTTCTCAATGACCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TEX33 PubMed Links

Gene Details

Gene
TEX33
Gene Name
testis expressed 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163857.1 834 Missense Mutation GAC,GGC D236G NP_001157329.1
NM_178552.3 834 Missense Mutation GAC,GGC D151G NP_848647.1
XM_011530165.2 834 Missense Mutation GAC,GGC D236G XP_011528467.1
XM_011530166.1 834 Missense Mutation GAC,GGC D151G XP_011528468.1

View Full Product Details