Product Details

SNP ID
rs146628737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:40679584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACAAGGTGGCAGGCGCTGGAGG[C/G]TGCCGCAGCCTGCGTGGGTGGAGGG
Phenotype
MIM: 601751
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MCHR1 PubMed Links

Gene Details

Gene
MCHR1
Gene Name
melanin concentrating hormone receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005297.3 407 Silent Mutation CTG,GTG L47V NP_005288.3

View Full Product Details