Product Details

SNP ID
rs146845551
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:16608454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTGACGCAGCTGTTGCTACCTC[A/G]CCTTTTCTTTTGTTGAGCTTAATCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TPTEP1 PubMed Links

Gene Details

Gene
TPTEP1
Gene Name
transmembrane phosphatase with tensin homology pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details