Product Details

SNP ID
rs146941567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38484017 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGACCTTTCCCTCAAACAGATGC[C/T]TTCAAGAGCTGCGAGAGAGTAGGGC
Phenotype
MIM: 608469
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DDX17 PubMed Links

Gene Details

Gene
DDX17
Gene Name
DEAD-box helicase 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098504.1 4217 UTR 3 NP_001091974.1
NM_006386.4 4217 UTR 3 NP_006377.2
Gene
KDELR3
Gene Name
KDEL endoplasmic reticulum protein retention receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006855.3 4217 Intron NP_006846.1
NM_016657.2 4217 Intron NP_057839.1

View Full Product Details