Product Details

SNP ID
rs147465498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:30579821 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAACGTAATCTCCTTCCTTCTC[A/G]GTCACAAAGGGTGAAAGGTGTGGGG
Phenotype
MIM: 602300 MIM: 605819
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GAL3ST1 PubMed Links

Gene Details

Gene
GAL3ST1
Gene Name
galactose-3-O-sulfotransferase 1
There are no transcripts associated with this gene.

Gene
PES1
Gene Name
pescadillo ribosomal biogenesis factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243225.1 1836 Intron NP_001230154.1
NM_001282327.1 1836 Silent Mutation NP_001269256.1
NM_001282328.1 1836 Silent Mutation NP_001269257.1
NM_014303.3 1836 Intron NP_055118.1
XM_006724173.1 1836 Silent Mutation XP_006724236.1
XM_017028678.1 1836 Silent Mutation XP_016884167.1

View Full Product Details