Product Details

SNP ID
rs147516607
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:35403316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCGACTACTTGTACAAGCAGCCA[A/G]CCGAGCACCTGCAGCTGGTGAGTGG
Phenotype
MIM: 141250 MIM: 602696
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HMOX1 PubMed Links

Gene Details

Gene
HMOX1
Gene Name
heme oxygenase 1
There are no transcripts associated with this gene.

Gene
MCM5
Gene Name
minichromosome maintenance complex component 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006739.3 426 Missense Mutation ACC,GCC T93A NP_006730.2
XM_006724242.3 426 Missense Mutation ACC,GCC T93A XP_006724305.1

View Full Product Details