Product Details

SNP ID
rs147546081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50446125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCAGCAGGTGAAGCCCTGGGC[C/T]TGGTGGGACTCAGGTTTGAATAGGA
Phenotype
MIM: 610877 MIM: 603560
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP6R2 PubMed Links

Gene Details

Gene
PPP6R2
Gene Name
protein phosphatase 6 regulatory subunit 2
There are no transcripts associated with this gene.

Gene
SBF1
Gene Name
SET binding factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002972.3 6930 UTR 3 NP_002963.2
XM_005261935.2 6930 Intron XP_005261992.1
XM_011530709.2 6930 Intron XP_011529011.1
XM_011530710.2 6930 Intron XP_011529012.1
XM_017028905.1 6930 Intron XP_016884394.1

View Full Product Details