Product Details
- SNP ID
-
rs147653734
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:49919793 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGCAGCGACTTCGAATGCAATCA[C/G]ATGCTAGAGGCGCAGGAGGAGCACC
- Phenotype
-
MIM: 607144
MIM: 607171
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ALG12
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9616388] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ALG12
- Gene Name
- ALG12, alpha-1,6-mannosyltransferase
There are no transcripts associated with this gene.
- Gene
- CRELD2
- Gene Name
- cysteine rich with EGF like domains 2
View Full Product Details