Product Details

SNP ID
rs147754758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:17138261 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTGAGGGCTGTTGCTGCCGTGT[A/G]CCCCCGGCCCCTAGTTCTGGCGCCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CECR5 PubMed Links

Gene Details

Gene
CECR5
Gene Name
cat eye syndrome chromosome region, candidate 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017829.5 1527 Silent Mutation GGC,GGT G314G NP_060299.4
NM_033070.2 1527 Silent Mutation GGC,GGT G344G NP_149061.1
XM_005261236.3 1527 Silent Mutation GGC,GGT G183G XP_005261293.1
XM_011546126.2 1527 Silent Mutation GGC,GGT G276G XP_011544428.1
XM_011546127.1 1527 Silent Mutation GGC,GGT G222G XP_011544429.1
XM_017028781.1 1527 Silent Mutation GGC,GGT G183G XP_016884270.1
XM_017028782.1 1527 Silent Mutation GGC,GGT G155G XP_016884271.1
XM_017028783.1 1527 Silent Mutation GGC,GGT G155G XP_016884272.1
XM_017028784.1 1527 Silent Mutation GGC,GGT G155G XP_016884273.1
Gene
LOC100996342
Gene Name
uncharacterized LOC100996342
There are no transcripts associated with this gene.

View Full Product Details