Product Details

SNP ID
rs147866417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:29791776 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGACCCCTTGCCTGAGAAGCAG[A/C]TAAATCTGTGGAAACTCTGAGTCTG
Phenotype
MIM: 614216
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ASCC2 PubMed Links

Gene Details

Gene
ASCC2
Gene Name
activating signal cointegrator 1 complex subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242906.1 Intron NP_001229835.1
NM_032204.4 Intron NP_115580.2
XM_005261775.2 Intron XP_005261832.1
XM_011530442.2 Intron XP_011528744.1
XM_011530443.2 Intron XP_011528745.1
XM_011530444.2 Intron XP_011528746.1
XM_011530445.2 Intron XP_011528747.1
XM_011530446.2 Intron XP_011528748.1
XM_011530448.2 Intron XP_011528750.1
XM_011530449.2 Intron XP_011528751.1
XM_011530450.2 Intron XP_011528752.1
XM_011530451.2 Intron XP_011528753.1
XM_011530452.2 Intron XP_011528754.1
XM_011530453.2 Intron XP_011528755.1
XM_011530454.2 Intron XP_011528756.1
XM_011530455.2 Intron XP_011528757.1
XM_017028991.1 Intron XP_016884480.1
XM_017028992.1 Intron XP_016884481.1
XM_017028993.1 Intron XP_016884482.1
XM_017028994.1 Intron XP_016884483.1
XM_017028995.1 Intron XP_016884484.1
XM_017028996.1 Intron XP_016884485.1
XM_017028997.1 Intron XP_016884486.1
XM_017028998.1 Intron XP_016884487.1
XM_017028999.1 Intron XP_016884488.1
XM_017029000.1 Intron XP_016884489.1
XM_017029001.1 Intron XP_016884490.1
XM_017029002.1 Intron XP_016884491.1
XM_017029003.1 Intron XP_016884492.1
XM_017029004.1 Intron XP_016884493.1
XM_017029005.1 Intron XP_016884494.1
XM_017029006.1 Intron XP_016884495.1
XM_017029007.1 Intron XP_016884496.1
XM_017029008.1 Intron XP_016884497.1
XM_017029009.1 Intron XP_016884498.1

View Full Product Details