Product Details

SNP ID
rs148469344
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:21572736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGCCAGACTCCAACCACTTCCA[A/G]AGTTTTCTTGGGAACGGCACATAGA
Phenotype
MIM: 603721
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UBE2L3 PubMed Links

Gene Details

Gene
UBE2L3
Gene Name
ubiquitin conjugating enzyme E2 L3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256355.1 Intron NP_001243284.1
NM_001256356.1 Intron NP_001243285.1
NM_003347.3 Intron NP_003338.1
XM_017028935.1 Intron XP_016884424.1

View Full Product Details