Product Details

SNP ID
rs148836234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:27749108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCTGAAGGCGGGGTTTGCTCCTC[A/G]TCAAGACTGCCCAGGCGAGAACCGC
Phenotype
MIM: 156100
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MN1 PubMed Links

Gene Details

Gene
MN1
Gene Name
MN1 proto-oncogene, transcriptional regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002430.2 6725 UTR 3 NP_002421.3

View Full Product Details