Product Details

SNP ID
rs149306114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:18157736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTGAGTCCTCGCAGTCCCCGGCA[A/G]ATCTTGAAGAAAAGAAGGAAGAAGA
Phenotype
MIM: 607057
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
USP18 PubMed Links

Gene Details

Gene
USP18
Gene Name
ubiquitin specific peptidase 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017414.3 443 Missense Mutation AAT,GAT N25D NP_059110.2
XM_006724074.2 443 UTR 5 XP_006724137.1

View Full Product Details