Product Details

SNP ID
rs149363275
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23753030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGCCGGTGGTGATCCTCCTCC[A/G]AGCGGTAGTAGAGGAGATATCGAGG
Phenotype
MIM: 605017 MIM: 194544
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C22orf15 PubMed Links

Gene Details

Gene
C22orf15
Gene Name
chromosome 22 open reading frame 15
There are no transcripts associated with this gene.

Gene
VPREB3
Gene Name
pre-B lymphocyte 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013378.2 298 Missense Mutation TCG,TTG S73L NP_037510.1
Gene
ZNF70
Gene Name
zinc finger protein 70
There are no transcripts associated with this gene.

View Full Product Details