Product Details

SNP ID
rs150043552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31496990 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCCTTAGCGTGCGCGGATCCAAA[C/T]CAGGGGTTTCCTCCTCCCGGCTCCG
Phenotype
MIM: 607445 MIM: 612765
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF4ENIF1 PubMed Links

Gene Details

Gene
EIF4ENIF1
Gene Name
eukaryotic translation initiation factor 4E nuclear import factor 1
There are no transcripts associated with this gene.

Gene
SFI1
Gene Name
SFI1 centrin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007467.2 Intron NP_001007468.1
NM_001258325.1 Intron NP_001245254.1
NM_001258326.1 Intron NP_001245255.1
NM_001258327.1 Intron NP_001245256.1
NM_014775.3 Intron NP_055590.2
XM_005261868.2 Intron XP_005261925.1
XM_006724390.2 Intron XP_006724453.1
XM_011530570.2 Intron XP_011528872.1
XM_011530571.2 Intron XP_011528873.1
XM_011530574.2 Intron XP_011528876.1
XM_011530575.2 Intron XP_011528877.1
XM_011530576.2 Intron XP_011528878.1
XM_011530577.2 Intron XP_011528879.1
XM_011530579.2 Intron XP_011528881.1
XM_011530580.2 Intron XP_011528882.1
XM_011530581.2 Intron XP_011528883.1
XM_011530582.2 Intron XP_011528884.1
XM_011530583.2 Intron XP_011528885.1
XM_011530587.2 Intron XP_011528889.1
XM_017029139.1 Intron XP_016884628.1
XM_017029140.1 Intron XP_016884629.1
XM_017029141.1 Intron XP_016884630.1
XM_017029142.1 Intron XP_016884631.1
XM_017029143.1 Intron XP_016884632.1
XM_017029144.1 Intron XP_016884633.1
XM_017029145.1 Intron XP_016884634.1
XM_017029146.1 Intron XP_016884635.1
XM_017029147.1 Intron XP_016884636.1
XM_017029148.1 Intron XP_016884637.1
XM_017029149.1 Intron XP_016884638.1
XM_017029150.1 Intron XP_016884639.1
XM_017029151.1 Intron XP_016884640.1
XM_017029152.1 Intron XP_016884641.1
XM_017029153.1 Intron XP_016884642.1
XM_017029154.1 Intron XP_016884643.1
XM_017029155.1 Intron XP_016884644.1
XM_017029156.1 Intron XP_016884645.1
XM_017029157.1 Intron XP_016884646.1
XM_017029158.1 Intron XP_016884647.1
XM_017029159.1 Intron XP_016884648.1
XM_017029160.1 Intron XP_016884649.1
XM_017029161.1 Intron XP_016884650.1
XM_017029162.1 Intron XP_016884651.1

View Full Product Details