Product Details
- SNP ID
-
rs150139304
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:31619805 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGCTGAAGTCATTGTAGGAGCCC[C/T]TGCTGTGCCTTGGGCTGTTTGTGTG
- Phenotype
-
MIM: 612770
MIM: 612765
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MIR7109
PubMed Links
Gene Details
- Gene
- MIR7109
- Gene Name
- microRNA 7109
There are no transcripts associated with this gene.
- Gene
- PISD
- Gene Name
- phosphatidylserine decarboxylase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001326411.1 |
1274 |
Missense Mutation |
AAG,AGG |
K346R |
NP_001313340.1 |
NM_001326412.1 |
1274 |
Missense Mutation |
AAG,AGG |
K325R |
NP_001313341.1 |
NM_001326413.1 |
1274 |
Missense Mutation |
AAG,AGG |
K325R |
NP_001313342.1 |
NM_001326414.1 |
1274 |
Missense Mutation |
AAG,AGG |
K325R |
NP_001313343.1 |
NM_001326415.1 |
1274 |
Missense Mutation |
AAG,AGG |
K312R |
NP_001313344.1 |
NM_001326416.1 |
1274 |
Missense Mutation |
AAG,AGG |
K312R |
NP_001313345.1 |
NM_001326417.1 |
1274 |
Missense Mutation |
AAG,AGG |
K312R |
NP_001313346.1 |
NM_001326418.1 |
1274 |
Missense Mutation |
AAG,AGG |
K286R |
NP_001313347.1 |
NM_001326419.1 |
1274 |
Missense Mutation |
AAG,AGG |
K274R |
NP_001313348.1 |
NM_001326420.1 |
1274 |
Missense Mutation |
AAG,AGG |
K248R |
NP_001313349.1 |
NM_001326421.1 |
1274 |
Silent Mutation |
CAA,CAG |
Q292Q |
NP_001313350.1 |
NM_014338.3 |
1274 |
Missense Mutation |
AAG,AGG |
K312R |
NP_055153.1 |
NM_178022.1 |
1274 |
Missense Mutation |
AAG,AGG |
K312R |
NP_821141.1 |
- Gene
- SFI1
- Gene Name
- SFI1 centrin binding protein
There are no transcripts associated with this gene.
View Full Product Details