Product Details

SNP ID
rs150178861
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:36141174 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCGTGGCCTGTGTATGGCTCACCT[C/G]CCCTGCTGGCTGCACTGGTCTGGGG
Phenotype
MIM: 607253
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
APOL3 PubMed Links

Gene Details

Gene
APOL3
Gene Name
apolipoprotein L3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014349.2 2206 UTR 3 NP_055164.1
NM_030644.1 2206 UTR 3 NP_085147.1
NM_145639.1 2206 UTR 3 NP_663614.1
NM_145640.2 2206 UTR 3 NP_663615.1
NM_145641.2 2206 UTR 3 NP_663616.1
NM_145642.2 2206 UTR 3 NP_663617.1
XM_006724324.1 2206 UTR 3 XP_006724387.1
XM_006724325.2 2206 UTR 3 XP_006724388.1
XM_017028946.1 2206 UTR 3 XP_016884435.1
XM_017028947.1 2206 UTR 3 XP_016884436.1
XM_017028948.1 2206 UTR 3 XP_016884437.1
XM_017028949.1 2206 UTR 3 XP_016884438.1
XM_017028950.1 2206 UTR 3 XP_016884439.1
XM_017028951.1 2206 UTR 3 XP_016884440.1
XM_017028952.1 2206 UTR 3 XP_016884441.1

View Full Product Details