Product Details

SNP ID
rs150278474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:50176988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCACAACTTCACGCGCGACCAGGC[A/G]CTGTACGCCCGCGGCTACTGCTGGA
Phenotype
MIM: 608421
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PANX2 PubMed Links

Gene Details

Gene
PANX2
Gene Name
pannexin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001160300.1 276 Silent Mutation GCA,GCG A92A NP_001153772.1
NM_052839.3 276 Silent Mutation GCA,GCG A92A NP_443071.2
Gene
TRABD
Gene Name
TraB domain containing
There are no transcripts associated with this gene.

View Full Product Details