Product Details

SNP ID
rs150521769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:31135889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTGTCGCCTCTGCTGAAGCCTC[C/T]GCAAGTGCCGGGCTGACACCCTGAT
Phenotype
MIM: 606481 MIM: 611651
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
INPP5J PubMed Links

Gene Details

Gene
INPP5J
Gene Name
inositol polyphosphate-5-phosphatase J
There are no transcripts associated with this gene.

Gene
PLA2G3
Gene Name
phospholipase A2 group III
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015715.4 1617 Missense Mutation CAG,CGG Q455R NP_056530.2
XM_011530204.1 1617 Missense Mutation CAG,CGG Q274R XP_011528506.1
XM_011530205.1 1617 Missense Mutation CAG,CGG Q274R XP_011528507.1

View Full Product Details