Product Details

SNP ID
rs150565069
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:20427540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGTTGAGTGTGTTATCCAGGTC[A/G]TGGTGGGCCACTGGGGAAGGATGAG
Phenotype
MIM: 613619
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCARF2 PubMed Links

Gene Details

Gene
SCARF2
Gene Name
scavenger receptor class F member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153334.6 1656 Silent Mutation NP_699165.3
NM_182895.4 1656 Silent Mutation NP_878315.2
XM_017029065.1 1656 Missense Mutation XP_016884554.1

View Full Product Details