Product Details

SNP ID
rs150895887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:18078506 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACCTTCTGGAGGAGGCGGCCGAC[C/T]TCCTGGTGGTGCACCTGGACTTCCG
Phenotype
MIM: 608666
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PEX26 PubMed Links

Gene Details

Gene
PEX26
Gene Name
peroxisomal biogenesis factor 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127649.2 339 Missense Mutation CTC,TTC L44F NP_001121121.1
NM_001199319.1 339 Missense Mutation CTC,TTC L44F NP_001186248.1
NM_017929.5 339 Missense Mutation CTC,TTC L44F NP_060399.1

View Full Product Details