Product Details
- SNP ID
-
rs150898918
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:24225284 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATGATGAGCTCCCCGCCAGCCCC[A/G]CCAATCACTAGCTTCGACCCCTGGG
- Phenotype
-
MIM: 137168
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GGT5
PubMed Links
Gene Details
- Gene
- GGT5
- Gene Name
- gamma-glutamyltransferase 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001099781.2 |
1785 |
Silent Mutation |
GGC,GGT |
G489G |
NP_001093251.1 |
NM_001099782.2 |
1785 |
Silent Mutation |
GGC,GGT |
G456G |
NP_001093252.1 |
NM_001302464.1 |
1785 |
Silent Mutation |
GGC,GGT |
G456G |
NP_001289393.1 |
NM_001302465.1 |
1785 |
Silent Mutation |
GGC,GGT |
G412G |
NP_001289394.1 |
NM_004121.3 |
1785 |
Silent Mutation |
GGC,GGT |
G488G |
NP_004112.2 |
XM_005261557.2 |
1785 |
Silent Mutation |
GGC,GGT |
G489G |
XP_005261614.1 |
XM_005261558.2 |
1785 |
Silent Mutation |
GGC,GGT |
G488G |
XP_005261615.1 |
XM_011530133.1 |
1785 |
Silent Mutation |
GGC,GGT |
G494G |
XP_011528435.1 |
XM_011530134.1 |
1785 |
Silent Mutation |
GGC,GGT |
G494G |
XP_011528436.1 |
XM_011530135.1 |
1785 |
Silent Mutation |
GGC,GGT |
G462G |
XP_011528437.1 |
XM_011530136.1 |
1785 |
Silent Mutation |
GGC,GGT |
G494G |
XP_011528438.1 |
XM_011530137.2 |
1785 |
Silent Mutation |
GGC,GGT |
G494G |
XP_011528439.1 |
XM_017028768.1 |
1785 |
Silent Mutation |
GGC,GGT |
G456G |
XP_016884257.1 |
XM_017028769.1 |
1785 |
Silent Mutation |
GGC,GGT |
G462G |
XP_016884258.1 |
View Full Product Details