Product Details

SNP ID
rs150924512
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23971570 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCAATCATAAAAAAGTCATGACC[A/G]TCCCTATCTTGCCAATCTGCCAGGA
Phenotype
MIM: 602750 MIM: 600437
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDT PubMed Links

Gene Details

Gene
DDT
Gene Name
D-dopachrome tautomerase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001084392.1 Intron NP_001077861.1
NM_001355.3 Intron NP_001346.1
Gene
DDTL
Gene Name
D-dopachrome tautomerase-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001084393.1 Intron NP_001077862.1
XM_011529816.2 Intron XP_011528118.1
Gene
GSTT2
Gene Name
glutathione S-transferase theta 2 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
LOC107985559
Gene Name
uncharacterized LOC107985559
There are no transcripts associated with this gene.

View Full Product Details