Product Details

SNP ID
rs151061359
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:35741371 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCAGCCAGCATCCTCTAACAAA[A/C]TCACGAAATGCCAGACTCCTCTGAC
Phenotype
MIM: 612149
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RBFOX2 PubMed Links

Gene Details

Gene
RBFOX2
Gene Name
RNA binding protein, fox-1 homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031695.2 4366 UTR 3 NP_001026865.1
NM_001082576.1 4366 UTR 3 NP_001076045.1
NM_001082577.1 4366 UTR 3 NP_001076046.1
NM_001082578.1 4366 UTR 3 NP_001076047.1
NM_001082579.1 4366 UTR 3 NP_001076048.1
NM_014309.2 4366 UTR 3 NP_055124.1
XM_005261428.2 4366 UTR 3 XP_005261485.1
XM_005261429.2 4366 UTR 3 XP_005261486.1
XM_005261430.2 4366 UTR 3 XP_005261487.1
XM_005261431.2 4366 UTR 3 XP_005261488.1
XM_005261432.2 4366 UTR 3 XP_005261489.1
XM_005261433.2 4366 UTR 3 XP_005261490.1
XM_005261435.1 4366 UTR 3 XP_005261492.1
XM_005261437.1 4366 UTR 3 XP_005261494.1
XM_006724185.2 4366 UTR 3 XP_006724248.1
XM_006724186.2 4366 UTR 3 XP_006724249.1
XM_006724187.2 4366 UTR 3 XP_006724250.1
XM_006724188.2 4366 UTR 3 XP_006724251.1
XM_006724189.3 4366 UTR 3 XP_006724252.1
XM_006724190.1 4366 UTR 3 XP_006724253.1
XM_006724191.1 4366 UTR 3 XP_006724254.1
XM_006724192.2 4366 UTR 3 XP_006724255.1
XM_006724193.3 4366 Intron XP_006724256.1
XM_006724194.2 4366 UTR 3 XP_006724257.1
XM_017028686.1 4366 UTR 3 XP_016884175.1
XM_017028687.1 4366 UTR 3 XP_016884176.1
XM_017028688.1 4366 UTR 3 XP_016884177.1
XM_017028689.1 4366 UTR 3 XP_016884178.1
XM_017028690.1 4366 UTR 3 XP_016884179.1
XM_017028691.1 4366 UTR 3 XP_016884180.1
XM_017028692.1 4366 UTR 3 XP_016884181.1
XM_017028693.1 4366 UTR 3 XP_016884182.1
XM_017028694.1 4366 UTR 3 XP_016884183.1
XM_017028695.1 4366 UTR 3 XP_016884184.1
XM_017028696.1 4366 UTR 3 XP_016884185.1
XM_017028697.1 4366 UTR 3 XP_016884186.1
XM_017028698.1 4366 UTR 3 XP_016884187.1

View Full Product Details