Product Details

SNP ID
rs151066753
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:46686828 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCTAACATTATTGCAATAGAGCC[A/G]CTAACGGGCCATTTTCTAAACTACA
Phenotype
MIM: 610307 MIM: 613691
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CERK PubMed Links

Gene Details

Gene
CERK
Gene Name
ceramide kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022766.5 2027 UTR 3 NP_073603.2
XM_017028909.1 2027 UTR 3 XP_016884398.1
Gene
GRAMD4
Gene Name
GRAM domain containing 4
There are no transcripts associated with this gene.

View Full Product Details